missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Nitrophthalic anhydride, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 168990250

 View more versions of this product

Catalog No. AC168990250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Nitrophthalic anhydride
641-70-3
100.0
Beige to Yellow
97%
C8H3NO5
25 g
3-nitrophthalic anhydride, 4-nitroisobenzofuran-1,3-dione, 1,3-isobenzofurandione, 4-nitro, phthalic anhydride, 3-nitro, 3-nitro phthalic anhydride, 3-nitrophthalic acid anhydride, 1,3-isobenzofurandione, nitro, 4-nitro-isobenzofuran-1,3-dione, 3-nitrophthalicanhydride, nitrophthalic anhydride
ROFZMKDROVBLNY-UHFFFAOYSA-N
4-nitro-2-benzofuran-1,3-dione
21631
97%
97%
96.0
160.0°C to 165.0°C
Authentic
Glass bottle
MFCD00005921
17, 486
Solubility in water: decomposes.
[O-][N+](=O)C1=CC=CC2=C1C(=O)OC2=O
193.11
193.12
Crystalline Powder

Chemical Identifiers

641-70-3
193.11
ROFZMKDROVBLNY-UHFFFAOYSA-N
21631
[O-][N+](=O)C1=CC=CC2=C1C(=O)OC2=O
C8H3NO5
MFCD00005921
3-nitrophthalic anhydride, 4-nitroisobenzofuran-1,3-dione, 1,3-isobenzofurandione, 4-nitro, phthalic anhydride, 3-nitro, 3-nitro phthalic anhydride, 3-nitrophthalic acid anhydride, 1,3-isobenzofurandione, nitro, 4-nitro-isobenzofuran-1,3-dione, 3-nitrophthalicanhydride, nitrophthalic anhydride
4-nitro-2-benzofuran-1,3-dione
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER

GHS Signal Word: Warning

EINECSNumber : 211-373-6

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only