missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Nitrophthalic anhydride, 97%, Thermo Scientific Chemicals

$222.57 - $222.57

Chemical Identifiers

CAS 641-70-3
Molecular Formula C8H3NO5
Molecular Weight (g/mol) 193.11
MDL Number MFCD00005921
InChI Key ROFZMKDROVBLNY-UHFFFAOYSA-N
Synonym 3-nitrophthalic anhydride, 4-nitroisobenzofuran-1,3-dione, 1,3-isobenzofurandione, 4-nitro, phthalic anhydride, 3-nitro, 3-nitro phthalic anhydride, 3-nitrophthalic acid anhydride, 1,3-isobenzofurandione, nitro, 4-nitro-isobenzofuran-1,3-dione, 3-nitrophthalicanhydride, nitrophthalic anhydride
PubChem CID 21631
IUPAC Name 4-nitro-2-benzofuran-1,3-dione
SMILES [O-][N+](=O)C1=CC=CC2=C1C(=O)OC2=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC168990250
SDS View Documents Promotion Details
Thermo Scientific Chemicals
168990250
25 g Glass bottle
Each for $222.57
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

641-70-3
193.11
ROFZMKDROVBLNY-UHFFFAOYSA-N
21631
[O-][N+](=O)C1=CC=CC2=C1C(=O)OC2=O
C8H3NO5
MFCD00005921
3-nitrophthalic anhydride, 4-nitroisobenzofuran-1,3-dione, 1,3-isobenzofurandione, 4-nitro, phthalic anhydride, 3-nitro, 3-nitro phthalic anhydride, 3-nitrophthalic acid anhydride, 1,3-isobenzofurandione, nitro, 4-nitro-isobenzofuran-1,3-dione, 3-nitrophthalicanhydride, nitrophthalic anhydride
4-nitro-2-benzofuran-1,3-dione

Specifications

641-70-3
100.0
Beige to Yellow
97%
C8H3NO5
25 g
3-nitrophthalic anhydride, 4-nitroisobenzofuran-1,3-dione, 1,3-isobenzofurandione, 4-nitro, phthalic anhydride, 3-nitro, 3-nitro phthalic anhydride, 3-nitrophthalic acid anhydride, 1,3-isobenzofurandione, nitro, 4-nitro-isobenzofuran-1,3-dione, 3-nitrophthalicanhydride, nitrophthalic anhydride
ROFZMKDROVBLNY-UHFFFAOYSA-N
4-nitro-2-benzofuran-1,3-dione
21631
97%
3-Nitrophthalic anhydride, 97%
96.0
160.0°C to 165.0°C
Authentic
Glass bottle
MFCD00005921
17, 486
Solubility in water: decomposes.
[O-][N+](=O)C1=CC=CC2=C1C(=O)OC2=O
193.11
193.12
Crystalline Powder
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER

GHS Signal Word: Warning

EINECSNumber : 211-373-6

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only