Learn More
3-Phenylazo-2,6-diaminopyridine hydrochloride, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2188606
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3-Phenylazo-2,6-diaminopyridine hydrochloride | |
∼240°C (decomposition) | |
MFCD00035347 | |
phenazopyridine hydrochloride, phenazopyridine hcl, urodine, pyridium, pyripyridium, bisteril, diridone, mallophene, phenazodine, pyrazodine | |
C1=CC=C(C=C1)N=NC2=C(N=C(C=C2)N)N.Cl | |
249.702 | |
CHEBI:71419 | |
≥98% |
136-40-3 | |
C11H12ClN5 | |
5 g | |
QQBPIHBUCMDKFG-UHFFFAOYSA-N | |
3-phenyldiazenylpyridine-2,6-diamine;hydrochloride | |
8691 | |
249.7 |
Chemical Identifiers
136-40-3 | |
249.702 | |
QQBPIHBUCMDKFG-UHFFFAOYSA-N | |
8691 | |
3-phenyldiazenylpyridine-2,6-diamine;hydrochloride |
C11H12ClN5 | |
MFCD00035347 | |
phenazopyridine hydrochloride, phenazopyridine hcl, urodine, pyridium, pyripyridium, bisteril, diridone, mallophene, phenazodine, pyrazodine | |
CHEBI:71419 | |
C1=CC=C(C=C1)N=NC2=C(N=C(C=C2)N)N.Cl |
Safety and Handling
GHS H Statement
H301-H334-H351-H317
Toxic if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing cancer.
May cause an allergic skin reaction.
P201-P202-P261-P264b-P270-P271-P272-P280-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P333+P313-P362-P501c
H302-H315-H317-H319-H335-H350
EINECSNumber : 205-243-8
RTECSNumber : US7875000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only