missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Sulfobenzoic acid monosodium salt, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2355614

 View more versions of this product

Catalog No. AAB2355614

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Sulfobenzoic acid monosodium salt
C7H5NaO5S
25 g
sodium 3-sulfobenzoate, 3-sulfobenzoic acid monosodium salt, benzoic acid, 3-sulfo-, monosodium salt, benzoic acid, 3-sulfo-, sodium salt 1:1, sodium hydrogen m-sulphonatobenzoate, sodium3-sulfobenzoate, sodium m-sulfobenzoate, acmc-209sh5, 3-sodiosulfo benzoic acid, ksc491e3l
C1=CC(=CC(=C1)S(=O)(=O)O)C(=O)[O-].[Na+]
224.162
224.17
17625-03-5
MFCD00066378
Hygroscopic
KQHKITXZJDOIOD-UHFFFAOYSA-M
sodium;3-sulfobenzoate
23668512
97%

Chemical Identifiers

17625-03-5
224.162
KQHKITXZJDOIOD-UHFFFAOYSA-M
sodium;3-sulfobenzoate
C7H5NaO5S
MFCD00066378
23668512
C1=CC(=CC(=C1)S(=O)(=O)O)C(=O)[O-].[Na+]
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 241-602-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only