missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Sulfobenzoic acid monosodium salt, 97%, Thermo Scientific Chemicals

$49.14 - $314.10

Chemical Identifiers

CAS 17625-03-5
Molecular Formula C7H5NaO5S
Molecular Weight (g/mol) 224.162
MDL Number MFCD00066378
InChI Key KQHKITXZJDOIOD-UHFFFAOYSA-M
PubChem CID 23668512
IUPAC Name sodium;3-sulfobenzoate
SMILES C1=CC(=CC(=C1)S(=O)(=O)O)C(=O)[O-].[Na+]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2355614
View Documents
Thermo Scientific Chemicals
B2355614
25 g
Each for $49.14
 
AAB2355622
View Documents
Thermo Scientific Chemicals
B2355622
100 g
Each for $97.33
 
AAB2355636
View Documents
Thermo Scientific Chemicals
B2355636
500 g
Each for $314.10
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

17625-03-5
224.162
KQHKITXZJDOIOD-UHFFFAOYSA-M
sodium;3-sulfobenzoate
C7H5NaO5S
MFCD00066378
23668512
C1=CC(=CC(=C1)S(=O)(=O)O)C(=O)[O-].[Na+]

Specifications

17625-03-5
MFCD00066378
Hygroscopic
KQHKITXZJDOIOD-UHFFFAOYSA-M
sodium;3-sulfobenzoate
23668512
97%
C7H5NaO5S
25 g
sodium 3-sulfobenzoate, 3-sulfobenzoic acid monosodium salt, benzoic acid, 3-sulfo-, monosodium salt, benzoic acid, 3-sulfo-, sodium salt 1:1, sodium hydrogen m-sulphonatobenzoate, sodium3-sulfobenzoate, sodium m-sulfobenzoate, acmc-209sh5, 3-sodiosulfo benzoic acid, ksc491e3l
C1=CC(=CC(=C1)S(=O)(=O)O)C(=O)[O-].[Na+]
224.162
224.17
3-Sulfobenzoic acid monosodium salt
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 241-602-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only