missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-(Trifluoromethyl)-5,6,7,8-tetrahydro-[1,2,4]triazolo[4,3-a]pyrazine Hydrochloride 98.0+%, TCI America™
Supplier: TCI America T29981G
Specifications
3-(Trifluoromethyl)-5,6,7,8-tetrahydro-[1,2,4]triazolo[4,3-a]pyrazine Hydrochloride | |
264°C | |
C6H8ClF3N4 | |
1 g | |
AQCSCRYRCRORET-UHFFFAOYSA-N | |
3-(trifluoromethyl)-5,6,7,8-tetrahydro-[1,2,4]triazolo[4,3-a]pyrazine;hydrochloride | |
11961371 | |
≥98.0% (HPLC) |
762240-92-6 | |
White | |
MFCD09817638 | |
3-trifluoromethyl-5,6,7,8-tetrahydro-1,2,4 triazolo 4,3-a pyrazine hydrochloride, 3-trifluoromethyl-5h,6h,7h,8h-1,2,4 triazolo 4,3-a pyrazine hydrochloride, 3-trifluoromethyl-5,6,7,8-tetrahydro-1,2,4-triazolo 4,3-a pyrazine hydrochloride, 3-trifluoromethyl-1,2,4-triazolo 4,3-a piperazine hydrochloride, 3-trifluoromethyl-5,6,7,8-tetrahydro-1,2,4 triazolo 4,3-a pyrazine hcl, 3-trifluoromethyl-5,6,7,8-tetrahydro-1,2,4-triazolo-4,3-a pyrazine hydrochloride, 3-trifluoromethyl-5,6,7,8-tetrahydro 1,2,4 triazolo 4,3-a pyrazine hydrochloride, 5,6,7,8-tetrahydro-3-trifluormethyl-4,3-a-1,2,4-triazolopyrazine hydrochloride, pubchem18203 | |
C1CN2C(=NN=C2C(F)(F)F)CN1.Cl | |
228.603 | |
228.60 | |
Crystalline Powder |
Chemical Identifiers
762240-92-6 | |
228.603 | |
AQCSCRYRCRORET-UHFFFAOYSA-N | |
11961371 | |
C1CN2C(=NN=C2C(F)(F)F)CN1.Cl |
C6H8ClF3N4 | |
MFCD09817638 | |
3-trifluoromethyl-5,6,7,8-tetrahydro-1,2,4 triazolo 4,3-a pyrazine hydrochloride, 3-trifluoromethyl-5h,6h,7h,8h-1,2,4 triazolo 4,3-a pyrazine hydrochloride, 3-trifluoromethyl-5,6,7,8-tetrahydro-1,2,4-triazolo 4,3-a pyrazine hydrochloride, 3-trifluoromethyl-1,2,4-triazolo 4,3-a piperazine hydrochloride, 3-trifluoromethyl-5,6,7,8-tetrahydro-1,2,4 triazolo 4,3-a pyrazine hcl, 3-trifluoromethyl-5,6,7,8-tetrahydro-1,2,4-triazolo-4,3-a pyrazine hydrochloride, 3-trifluoromethyl-5,6,7,8-tetrahydro 1,2,4 triazolo 4,3-a pyrazine hydrochloride, 5,6,7,8-tetrahydro-3-trifluormethyl-4,3-a-1,2,4-triazolopyrazine hydrochloride, pubchem18203 | |
3-(trifluoromethyl)-5,6,7,8-tetrahydro-[1,2,4]triazolo[4,3-a]pyrazine;hydrochloride |
Safety and Handling
TSCA : No