missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(Trifluoromethyl)-5,6,7,8-tetrahydro-[1,2,4]triazolo[4,3-a]pyrazine Hydrochloride 98.0+%, TCI America™

$103.81 - $304.49

Chemical Identifiers

CAS 762240-92-6
Molecular Formula C6H8ClF3N4
Molecular Weight (g/mol) 228.603
MDL Number MFCD09817638
InChI Key AQCSCRYRCRORET-UHFFFAOYSA-N
Synonym 3-trifluoromethyl-5,6,7,8-tetrahydro-1,2,4 triazolo 4,3-a pyrazine hydrochloride, 3-trifluoromethyl-5h,6h,7h,8h-1,2,4 triazolo 4,3-a pyrazine hydrochloride, 3-trifluoromethyl-5,6,7,8-tetrahydro-1,2,4-triazolo 4,3-a pyrazine hydrochloride, 3-trifluoromethyl-1,2,4-triazolo 4,3-a piperazine hydrochloride, 3-trifluoromethyl-5,6,7,8-tetrahydro-1,2,4 triazolo 4,3-a pyrazine hcl, 3-trifluoromethyl-5,6,7,8-tetrahydro-1,2,4-triazolo-4,3-a pyrazine hydrochloride, 3-trifluoromethyl-5,6,7,8-tetrahydro 1,2,4 triazolo 4,3-a pyrazine hydrochloride, 5,6,7,8-tetrahydro-3-trifluormethyl-4,3-a-1,2,4-triazolopyrazine hydrochloride, pubchem18203
PubChem CID 11961371
IUPAC Name 3-(trifluoromethyl)-5,6,7,8-tetrahydro-[1,2,4]triazolo[4,3-a]pyrazine;hydrochloride
SMILES C1CN2C(=NN=C2C(F)(F)F)CN1.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
T29981G
View Documents
TCI America
T29981G
1 g
Each for $103.81
 
T29985G
View Documents
TCI America
T29985G
5 g
Each for $304.49
 
Description

Description

Specifications

Chemical Identifiers

762240-92-6
228.603
AQCSCRYRCRORET-UHFFFAOYSA-N
11961371
C1CN2C(=NN=C2C(F)(F)F)CN1.Cl
C6H8ClF3N4
MFCD09817638
3-trifluoromethyl-5,6,7,8-tetrahydro-1,2,4 triazolo 4,3-a pyrazine hydrochloride, 3-trifluoromethyl-5h,6h,7h,8h-1,2,4 triazolo 4,3-a pyrazine hydrochloride, 3-trifluoromethyl-5,6,7,8-tetrahydro-1,2,4-triazolo 4,3-a pyrazine hydrochloride, 3-trifluoromethyl-1,2,4-triazolo 4,3-a piperazine hydrochloride, 3-trifluoromethyl-5,6,7,8-tetrahydro-1,2,4 triazolo 4,3-a pyrazine hcl, 3-trifluoromethyl-5,6,7,8-tetrahydro-1,2,4-triazolo-4,3-a pyrazine hydrochloride, 3-trifluoromethyl-5,6,7,8-tetrahydro 1,2,4 triazolo 4,3-a pyrazine hydrochloride, 5,6,7,8-tetrahydro-3-trifluormethyl-4,3-a-1,2,4-triazolopyrazine hydrochloride, pubchem18203
3-(trifluoromethyl)-5,6,7,8-tetrahydro-[1,2,4]triazolo[4,3-a]pyrazine;hydrochloride

Specifications

762240-92-6
White
MFCD09817638
3-trifluoromethyl-5,6,7,8-tetrahydro-1,2,4 triazolo 4,3-a pyrazine hydrochloride, 3-trifluoromethyl-5h,6h,7h,8h-1,2,4 triazolo 4,3-a pyrazine hydrochloride, 3-trifluoromethyl-5,6,7,8-tetrahydro-1,2,4-triazolo 4,3-a pyrazine hydrochloride, 3-trifluoromethyl-1,2,4-triazolo 4,3-a piperazine hydrochloride, 3-trifluoromethyl-5,6,7,8-tetrahydro-1,2,4 triazolo 4,3-a pyrazine hcl, 3-trifluoromethyl-5,6,7,8-tetrahydro-1,2,4-triazolo-4,3-a pyrazine hydrochloride, 3-trifluoromethyl-5,6,7,8-tetrahydro 1,2,4 triazolo 4,3-a pyrazine hydrochloride, 5,6,7,8-tetrahydro-3-trifluormethyl-4,3-a-1,2,4-triazolopyrazine hydrochloride, pubchem18203
C1CN2C(=NN=C2C(F)(F)F)CN1.Cl
228.603
228.60
Crystalline Powder
264°C
C6H8ClF3N4
1 g
AQCSCRYRCRORET-UHFFFAOYSA-N
3-(trifluoromethyl)-5,6,7,8-tetrahydro-[1,2,4]triazolo[4,3-a]pyrazine;hydrochloride
11961371
≥98.0% (HPLC)
3-(Trifluoromethyl)-5,6,7,8-tetrahydro-[1,2,4]triazolo[4,3-a]pyrazine Hydrochloride
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'