missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(Trifluoromethyl)-5,6,7,8-tetrahydro-[1,2,4]triazolo[4,3-a]pyrazine Hydrochloride 98.0+%, TCI America™

Supplier:  TCI America T29985G

 View more versions of this product

Catalog No. T29985G


Explore available promotions

Description

Description

Specifications

Specifications

3-(Trifluoromethyl)-5,6,7,8-tetrahydro-[1,2,4]triazolo[4,3-a]pyrazine Hydrochloride
264°C
C6H8ClF3N4
5 g
AQCSCRYRCRORET-UHFFFAOYSA-N
3-(trifluoromethyl)-5,6,7,8-tetrahydro-[1,2,4]triazolo[4,3-a]pyrazine;hydrochloride
11961371
≥98.0% (HPLC)
762240-92-6
White
MFCD09817638
3-trifluoromethyl-5,6,7,8-tetrahydro-1,2,4 triazolo 4,3-a pyrazine hydrochloride, 3-trifluoromethyl-5h,6h,7h,8h-1,2,4 triazolo 4,3-a pyrazine hydrochloride, 3-trifluoromethyl-5,6,7,8-tetrahydro-1,2,4-triazolo 4,3-a pyrazine hydrochloride, 3-trifluoromethyl-1,2,4-triazolo 4,3-a piperazine hydrochloride, 3-trifluoromethyl-5,6,7,8-tetrahydro-1,2,4 triazolo 4,3-a pyrazine hcl, 3-trifluoromethyl-5,6,7,8-tetrahydro-1,2,4-triazolo-4,3-a pyrazine hydrochloride, 3-trifluoromethyl-5,6,7,8-tetrahydro 1,2,4 triazolo 4,3-a pyrazine hydrochloride, 5,6,7,8-tetrahydro-3-trifluormethyl-4,3-a-1,2,4-triazolopyrazine hydrochloride, pubchem18203
C1CN2C(=NN=C2C(F)(F)F)CN1.Cl
228.603
228.60
Crystalline Powder

Chemical Identifiers

762240-92-6
228.603
AQCSCRYRCRORET-UHFFFAOYSA-N
11961371
C1CN2C(=NN=C2C(F)(F)F)CN1.Cl
C6H8ClF3N4
MFCD09817638
3-trifluoromethyl-5,6,7,8-tetrahydro-1,2,4 triazolo 4,3-a pyrazine hydrochloride, 3-trifluoromethyl-5h,6h,7h,8h-1,2,4 triazolo 4,3-a pyrazine hydrochloride, 3-trifluoromethyl-5,6,7,8-tetrahydro-1,2,4-triazolo 4,3-a pyrazine hydrochloride, 3-trifluoromethyl-1,2,4-triazolo 4,3-a piperazine hydrochloride, 3-trifluoromethyl-5,6,7,8-tetrahydro-1,2,4 triazolo 4,3-a pyrazine hcl, 3-trifluoromethyl-5,6,7,8-tetrahydro-1,2,4-triazolo-4,3-a pyrazine hydrochloride, 3-trifluoromethyl-5,6,7,8-tetrahydro 1,2,4 triazolo 4,3-a pyrazine hydrochloride, 5,6,7,8-tetrahydro-3-trifluormethyl-4,3-a-1,2,4-triazolopyrazine hydrochloride, pubchem18203
3-(trifluoromethyl)-5,6,7,8-tetrahydro-[1,2,4]triazolo[4,3-a]pyrazine;hydrochloride
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions