Learn More
3-(Trifluoromethyl)phenyl isothiocyanate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1164614
Description
Used as pharamaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsUsed as pharamaceutical intermediate.
Solubility
Hydrolyzes in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to moisture.
Specifications
3-(Trifluoromethyl)phenyl isothiocyanate | |
1.335 | |
95°C (203°F) | |
C8H4F3NS | |
MFCD00004806 | |
UN2922 | |
Moisture sensitive | |
Hydrolyzes in water. | |
C1=CC(=CC(=C1)N=C=S)C(F)(F)F | |
203.182 | |
203.19 |
1840-19-3 | |
208°C | |
Pungent | |
1.5547 | |
25 g | |
781205 | |
3-trifluoromethyl phenyl isothiocyanate, 1-isothiocyanato-3-trifluoromethyl benzene, 3-trifluoromethyl phenylisothiocyanate, benzene, 1-isothiocyanato-3-trifluoromethyl, m-trifluoromethyl phenyl isothiocyanate, alpha,alpha,alpha-trifluoro-m-tolyl isothiocyanate, 3-trifluoromethylphenyl isothiocyanate, 1-isothiocyanato-3-trifluoromethyl-benzene, benzene,1-isothiocyanato-3-trifluoromethyl | |
GFEPANUKFYVALF-UHFFFAOYSA-N | |
1-isothiocyanato-3-(trifluoromethyl)benzene | |
137232 | |
98% |
Chemical Identifiers
1840-19-3 | |
203.182 | |
GFEPANUKFYVALF-UHFFFAOYSA-N | |
137232 | |
C1=CC(=CC(=C1)N=C=S)C(F)(F)F |
C8H4F3NS | |
MFCD00004806 | |
3-trifluoromethyl phenyl isothiocyanate, 1-isothiocyanato-3-trifluoromethyl benzene, 3-trifluoromethyl phenylisothiocyanate, benzene, 1-isothiocyanato-3-trifluoromethyl, m-trifluoromethyl phenyl isothiocyanate, alpha,alpha,alpha-trifluoro-m-tolyl isothiocyanate, 3-trifluoromethylphenyl isothiocyanate, 1-isothiocyanato-3-trifluoromethyl-benzene, benzene,1-isothiocyanato-3-trifluoromethyl | |
1-isothiocyanato-3-(trifluoromethyl)benzene |
Safety and Handling
GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: II
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only