Learn More
3-(Trifluoromethyl)phenyl isothiocyanate, 98%, Thermo Scientific Chemicals
$46.36 - $328.84
Chemical Identifiers
CAS | 1840-19-3 |
---|---|
Molecular Formula | C8H4F3NS |
Molecular Weight (g/mol) | 203.182 |
MDL Number | MFCD00004806 |
InChI Key | GFEPANUKFYVALF-UHFFFAOYSA-N |
Synonym | 3-trifluoromethyl phenyl isothiocyanate, 1-isothiocyanato-3-trifluoromethyl benzene, 3-trifluoromethyl phenylisothiocyanate, benzene, 1-isothiocyanato-3-trifluoromethyl, m-trifluoromethyl phenyl isothiocyanate, alpha,alpha,alpha-trifluoro-m-tolyl isothiocyanate, 3-trifluoromethylphenyl isothiocyanate, 1-isothiocyanato-3-trifluoromethyl-benzene, benzene,1-isothiocyanato-3-trifluoromethyl |
PubChem CID | 137232 |
IUPAC Name | 1-isothiocyanato-3-(trifluoromethyl)benzene |
SMILES | C1=CC(=CC(=C1)N=C=S)C(F)(F)F |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1164603
|
Thermo Scientific Chemicals
L1164603 |
1 g |
Each for $46.36
|
|
|||||
AAL1164606
|
Thermo Scientific Chemicals
L1164606 |
5 g |
Each for $101.60
|
|
|||||
AAL1164614
|
Thermo Scientific Chemicals
L1164614 |
25 g |
Each for $328.84
|
|
|||||
Description
Used as pharamaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsUsed as pharamaceutical intermediate.
Solubility
Hydrolyzes in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to moisture.
Chemical Identifiers
1840-19-3 | |
203.182 | |
GFEPANUKFYVALF-UHFFFAOYSA-N | |
137232 | |
C1=CC(=CC(=C1)N=C=S)C(F)(F)F |
C8H4F3NS | |
MFCD00004806 | |
3-trifluoromethyl phenyl isothiocyanate, 1-isothiocyanato-3-trifluoromethyl benzene, 3-trifluoromethyl phenylisothiocyanate, benzene, 1-isothiocyanato-3-trifluoromethyl, m-trifluoromethyl phenyl isothiocyanate, alpha,alpha,alpha-trifluoro-m-tolyl isothiocyanate, 3-trifluoromethylphenyl isothiocyanate, 1-isothiocyanato-3-trifluoromethyl-benzene, benzene,1-isothiocyanato-3-trifluoromethyl | |
1-isothiocyanato-3-(trifluoromethyl)benzene |
Specifications
1840-19-3 | |
208°C | |
Pungent | |
1.5547 | |
1 g | |
781205 | |
3-trifluoromethyl phenyl isothiocyanate, 1-isothiocyanato-3-trifluoromethyl benzene, 3-trifluoromethyl phenylisothiocyanate, benzene, 1-isothiocyanato-3-trifluoromethyl, m-trifluoromethyl phenyl isothiocyanate, alpha,alpha,alpha-trifluoro-m-tolyl isothiocyanate, 3-trifluoromethylphenyl isothiocyanate, 1-isothiocyanato-3-trifluoromethyl-benzene, benzene,1-isothiocyanato-3-trifluoromethyl | |
GFEPANUKFYVALF-UHFFFAOYSA-N | |
1-isothiocyanato-3-(trifluoromethyl)benzene | |
137232 | |
98% |
1.335 | |
95°C (203°F) | |
C8H4F3NS | |
MFCD00004806 | |
UN2922 | |
Moisture sensitive | |
Hydrolyzes in water. | |
C1=CC(=CC(=C1)N=C=S)C(F)(F)F | |
203.182 | |
203.19 | |
3-(Trifluoromethyl)phenyl isothiocyanate |
Safety and Handling
GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: II
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only