missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(Trifluoromethylthio)benzoic acid, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1997506

 View more versions of this product

Catalog No. AAA1997506


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-(Trifluoromethylthio)benzoic acid
71°C to 74°C
MFCD00236337
3-trifluoromethylthio benzoic acid, 3-trifluoromethyl thio benzoic acid, 3-trifluoromethyl sulfanyl benzoic acid, 3-trifluoromethylsulfanyl benzoic acid, 3-trifluoromethylsulfanyl-benzoic acid, benzoic acid, 3-trifluoromethyl thio, 3-trifluoromethyl sulphanyl benzoic acid, 3-trifluoromethylthio benzoicacid, 3-carboxyphenyl trifluoromethyl sulphide
[O-]C(=O)C1=CC=CC(SC(F)(F)F)=C1
221.17
222.18
946-65-6
C8H4F3O2S
5 g
IVGAPIVNQABETQ-UHFFFAOYSA-M
3-(trifluoromethylsulfanyl)benzoic acid
688330
97%

Chemical Identifiers

946-65-6
221.17
IVGAPIVNQABETQ-UHFFFAOYSA-M
688330
[O-]C(=O)C1=CC=CC(SC(F)(F)F)=C1
C8H4F3O2S
MFCD00236337
3-trifluoromethylthio benzoic acid, 3-trifluoromethyl thio benzoic acid, 3-trifluoromethyl sulfanyl benzoic acid, 3-trifluoromethylsulfanyl benzoic acid, 3-trifluoromethylsulfanyl-benzoic acid, benzoic acid, 3-trifluoromethyl thio, 3-trifluoromethyl sulphanyl benzoic acid, 3-trifluoromethylthio benzoicacid, 3-carboxyphenyl trifluoromethyl sulphide
3-(trifluoromethylsulfanyl)benzoic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only