missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(Trifluoromethylthio)benzoic acid, 97%, Thermo Scientific Chemicals

$113.85 - $424.92

Chemical Identifiers

CAS 946-65-6
Molecular Formula C8H4F3O2S
Molecular Weight (g/mol) 221.17
MDL Number MFCD00236337
InChI Key IVGAPIVNQABETQ-UHFFFAOYSA-M
Synonym 3-trifluoromethylthio benzoic acid, 3-trifluoromethyl thio benzoic acid, 3-trifluoromethyl sulfanyl benzoic acid, 3-trifluoromethylsulfanyl benzoic acid, 3-trifluoromethylsulfanyl-benzoic acid, benzoic acid, 3-trifluoromethyl thio, 3-trifluoromethyl sulphanyl benzoic acid, 3-trifluoromethylthio benzoicacid, 3-carboxyphenyl trifluoromethyl sulphide
PubChem CID 688330
IUPAC Name 3-(trifluoromethylsulfanyl)benzoic acid
SMILES [O-]C(=O)C1=CC=CC(SC(F)(F)F)=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1997503
View Documents
Thermo Scientific Chemicals
A1997503
1 g
Each for $113.85
 
AAA1997506
View Documents
Thermo Scientific Chemicals
A1997506
5 g
Each for $424.92
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

946-65-6
221.17
IVGAPIVNQABETQ-UHFFFAOYSA-M
688330
[O-]C(=O)C1=CC=CC(SC(F)(F)F)=C1
C8H4F3O2S
MFCD00236337
3-trifluoromethylthio benzoic acid, 3-trifluoromethyl thio benzoic acid, 3-trifluoromethyl sulfanyl benzoic acid, 3-trifluoromethylsulfanyl benzoic acid, 3-trifluoromethylsulfanyl-benzoic acid, benzoic acid, 3-trifluoromethyl thio, 3-trifluoromethyl sulphanyl benzoic acid, 3-trifluoromethylthio benzoicacid, 3-carboxyphenyl trifluoromethyl sulphide
3-(trifluoromethylsulfanyl)benzoic acid

Specifications

946-65-6
C8H4F3O2S
1 g
IVGAPIVNQABETQ-UHFFFAOYSA-M
3-(trifluoromethylsulfanyl)benzoic acid
688330
97%
71°C to 74°C
MFCD00236337
3-trifluoromethylthio benzoic acid, 3-trifluoromethyl thio benzoic acid, 3-trifluoromethyl sulfanyl benzoic acid, 3-trifluoromethylsulfanyl benzoic acid, 3-trifluoromethylsulfanyl-benzoic acid, benzoic acid, 3-trifluoromethyl thio, 3-trifluoromethyl sulphanyl benzoic acid, 3-trifluoromethylthio benzoicacid, 3-carboxyphenyl trifluoromethyl sulphide
[O-]C(=O)C1=CC=CC(SC(F)(F)F)=C1
221.17
222.18
3-(Trifluoromethylthio)benzoic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only