missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(Trimethoxysilyl)propyl methacrylate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 216550050

 View more versions of this product

Catalog No. AC216550050


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-(Trimethoxysilyl)propyl methacrylate
97.5
1.0400g/mL
100°C
97.5% min. (GC)
C10H20O5Si
H2C=C(CH3)CO2(CH2)3Si(OCH3)3
5 mL
3-trimethoxysilyl propyl methacrylate, 3-methacryloxypropyltrimethoxysilane, methacryloxypropyltrimethoxysilane, dynasylan memo, silicone a-174, union carbide a-174, mops-m, silane a-174, nuca 174, 2-propenoic acid, 2-methyl-, 3-trimethoxysilyl propyl ester
XDLMVUHYZWKMMD-UHFFFAOYSA-N
3-trimethoxysilylpropyl 2-methylprop-2-enoate
17318
248.35
Liquid
2530-85-0
100.0
190.0°C
Authentic
Glass bottle
1.4300 to 1.4320
MFCD00008593
1.04
Solubility in water: insoluble. Other solubilities: soluble in acetone,benzene,ether,methanol and,hydrocarbons
CC(=C)C(=O)OCCC[Si](OC)(OC)OC
248.35
2.5 mm2/s (25°C)
98%

Chemical Identifiers

2530-85-0
248.35
XDLMVUHYZWKMMD-UHFFFAOYSA-N
17318
CC(=C)C(=O)OCCC[Si](OC)(OC)OC
C10H20O5Si
MFCD00008593
3-trimethoxysilyl propyl methacrylate, 3-methacryloxypropyltrimethoxysilane, methacryloxypropyltrimethoxysilane, dynasylan memo, silicone a-174, union carbide a-174, mops-m, silane a-174, nuca 174, 2-propenoic acid, 2-methyl-, 3-trimethoxysilyl propyl ester
3-trimethoxysilylpropyl 2-methylprop-2-enoate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

EINECSNumber : 219-785-8

Recommended Storage : Product may darken on storage

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only