Learn More
3-(Trimethoxysilyl)propyl methacrylate, 98%
CAS: 2530-85-0 | C10H20O5Si | 248.35 g/mol
$85.84 - $184.12
Chemical Identifiers
| CAS | 2530-85-0 |
|---|---|
| Molecular Formula | C10H20O5Si |
| Molecular Weight (g/mol) | 248.35 |
| MDL Number | MFCD00008593 |
| InChI Key | XDLMVUHYZWKMMD-UHFFFAOYSA-N |
| Synonym | 3-trimethoxysilyl propyl methacrylate, 3-methacryloxypropyltrimethoxysilane, methacryloxypropyltrimethoxysilane, dynasylan memo, silicone a-174, union carbide a-174, mops-m, silane a-174, nuca 174, 2-propenoic acid, 2-methyl-, 3-trimethoxysilyl propyl ester |
| PubChem CID | 17318 |
| IUPAC Name | 3-trimethoxysilylpropyl 2-methylprop-2-enoate |
| SMILES | CC(=C)C(=O)OCCC[Si](OC)(OC)OC |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC216550050
|
Thermo Scientific Chemicals
216550050 |
5 mL | Glass bottle |
Each for $85.84
|
|
||||
|
AC216551000
|
Thermo Scientific Chemicals
216551000 |
100 mL | Glass bottle |
Each for $114.78
|
|
||||
|
AC216555000
|
Thermo Scientific Chemicals
216555000 |
500 mL | Glass bottle |
Each for $184.12
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 2530-85-0 | |
| 248.35 | |
| XDLMVUHYZWKMMD-UHFFFAOYSA-N | |
| 17318 | |
| CC(=C)C(=O)OCCC[Si](OC)(OC)OC |
| C10H20O5Si | |
| MFCD00008593 | |
| 3-trimethoxysilyl propyl methacrylate, 3-methacryloxypropyltrimethoxysilane, methacryloxypropyltrimethoxysilane, dynasylan memo, silicone a-174, union carbide a-174, mops-m, silane a-174, nuca 174, 2-propenoic acid, 2-methyl-, 3-trimethoxysilyl propyl ester | |
| 3-trimethoxysilylpropyl 2-methylprop-2-enoate |
Specifications
| 2530-85-0 | |
| 100.0 | |
| 190.0°C | |
| Authentic | |
| Glass bottle | |
| 1.4300 to 1.4320 | |
| MFCD00008593 | |
| 1.04 | |
| Solubility in water: insoluble. Other solubilities: soluble in acetone,benzene,ether,methanol and,hydrocarbons | |
| CC(=C)C(=O)OCCC[Si](OC)(OC)OC | |
| 248.35 | |
| 2.5 mm2/s (25°C) | |
| 98% | |
| 3-(Trimethoxysilyl)propyl methacrylate |
| 97.5 | |
| 1.0400g/mL | |
| 100°C | |
| 97.5% min. (GC) | |
| C10H20O5Si | |
| H2C=C(CH3)CO2(CH2)3Si(OCH3)3 | |
| 5 mL | |
| 3-trimethoxysilyl propyl methacrylate, 3-methacryloxypropyltrimethoxysilane, methacryloxypropyltrimethoxysilane, dynasylan memo, silicone a-174, union carbide a-174, mops-m, silane a-174, nuca 174, 2-propenoic acid, 2-methyl-, 3-trimethoxysilyl propyl ester | |
| XDLMVUHYZWKMMD-UHFFFAOYSA-N | |
| 3-trimethoxysilylpropyl 2-methylprop-2-enoate | |
| 17318 | |
| 248.35 | |
| Liquid |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
EINECSNumber : 219-785-8
Recommended Storage : Product may darken on storage
RUO – Research Use Only