missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Biotix™ 384-Square Deep Well Microplates

Made from medical-grade virgin polypropylene.

Supplier:  Biotix™ 63300112

 View more versions of this product

Catalog No. 12-111-080



Explore available promotions

Description

Description

  • Withstands centrifugation up to 6000 RCF
  • DNase, RNase and endotoxin (pyrogen) free
Specifications

Specifications

No
Without Lid
Non Binding
Polypropylene
384
Non-treated
10
22 mm
Standard
16 x 24
Clear
190 μL
Non-sterile
Square
85.48 mm
Videos
SDS
Documents

Documents

Promotions

Promotions