missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3B Scientific™ Anatomy Posters

Visual aids to enhance anatomy lessons.

Supplier:  3B Scientific™ VR1353L

 View more versions of this product

Catalog No. S90765


Explore available promotions

Description

Description

  • Printed on premium glossy (200g) paper
  • With two-sided lamination (125 micron) and metal eyelets
  • Dimensions: 20W × 26 in.H (50.8 × 66.0cm)

Specifications

Specifications

Science
26 in.
20 in.
The Vascular System Chart
66 cm
50.8 cm
Videos
SDS
Documents

Documents

Promotions

Promotions