3M™ 6000 Series Reusable Full Facepiece Respirator
Requires limited maintenance time—reducing need for expensive repair programs
Supplier:  3M™ 7000002029
 View more versions of this product
Catalog No. 18-999-4555
Explore available promotions
Requires : Compatible cartridges and/or filters (sold separately). Compatible with Series 2000 and 7093 filters and Series 6000 cartridges.
Description
Description
  • No retainers or extra parts needed to attach filters, cartridges or dual airline
  • Soft, lightweight facepiece material and easy-to-use headstraps
  • Large lens with scratch-resistant hardcoat
  • 3M Catalog# 6700 , 3M Catalog# 6800 , 3M Catalog# 6900
These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product. Misuse may result in sickness or death.
Certifications
NIOSH approved for 3M Negative Pressure Air-Purifying Respirators, Positive-Pressure Dual Airline Respirators, and some 3M PAPRs (with DIN adapter).
Compliance
NIOSH approved for 3M Negative Pressure Air-Purifying Respirators, Positive-Pressure Dual Airline Respirators, and some 3M PAPRS (with DIN adapter).
Specifications
Specifications
NIOSH, ANSI Z87.1-2010
Silicone, Thermoplastic Elastomer
Small
3M™ cartridge 6000 series, filter 2000, 2200, 7000 or 5000 series or 3M™ dual airline system
6700
Bayonet
Full Facepiece Respirator
Assembly, cleaning, demolition, facility maintenance, grinding, machine operation, painting, remediation, sanding, welding
Large lens with scratch-resistant hard coating
4-point
Videos
SDS
Documents
Documents
Accessories