3M™ Breathe Easy™ Filters, Cartridges, and Canisters
Used for protection against a combination of organic vapors, hydrogen fluoride, sulfur dioxide and/or high efficiency particulates.
Supplier:  3M™ 7000131720
 View more versions of this product
Catalog No. 17-987-272G
Description
Description
  • NIOSH-approved P3 particulate cartridges used for Breathe Easy PAPR systems
  • High efficiency (HE) respiratory protection against particulates
  • 3M Catalog# 450-00-01R12, 453-00-01R06, 453-02-01R06, 453-03-01R06, 453-07-01R06
Specifications
Specifications
Organic Vapor, Chlorine, Hydrogen Chloride, Sulfur Dioxide Cartridge
Black
Combination Cartridge/Filter
453-03-01R06
NIOSH, MSHA
High Efficiency (HE)
Breathe Easy™ PAPR Systems
6/Pk.