3M™ Breathe Easy™ Filters, Cartridges, and Canisters
Used for protection against a combination of organic vapors, hydrogen fluoride, sulfur dioxide and/or high efficiency particulates.
$171.74 - $525.79
Specifications
Certifications/Compliance | NIOSH, MSHA |
---|---|
Filter Type | High Efficiency (HE) |
For Use With (Equipment) | Breathe Easy™ PAPR Systems |
Catalog Number | Mfr. No. | Cartridge Type | Color | Product Type | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Cartridge Type | Color | Product Type | Price | Quantity | |||
17-987-272E
|
3M™
7000131700 |
Filter Cartridge | Magenta | HE Filter |
Case of 12 Each for $525.79
|
|
|||
17-987-272G
|
3M™
7000131720 |
Organic Vapor, Chlorine, Hydrogen Chloride, Sulfur Dioxide Cartridge | Black | Combination Cartridge/Filter |
Case of 6 Each for $171.74
|
|
|||
17-987-272J
|
3M™
7000126358 |
Ammonia, Methylamine Cartridge | Magenta/Green | Combination Cartridge/Filter |
Pack of 6 Each for $409.11
|
|
|||
Description
- NIOSH-approved P3 particulate cartridges used for Breathe Easy PAPR systems
- High efficiency (HE) respiratory protection against particulates
- 3M Catalog# 450-00-01R12, 453-00-01R06, 453-02-01R06, 453-03-01R06, 453-07-01R06
Specifications
NIOSH, MSHA | |
High Efficiency (HE) | |
Breathe Easy™ PAPR Systems |