3M™ Breathe Easy™ Filters, Cartridges, and Canisters
Used for protection against a combination of organic vapors, hydrogen fluoride, sulfur dioxide and/or high efficiency particulates.
Supplier: 3M™ 7000126358
Description
- NIOSH-approved P3 particulate cartridges used for Breathe Easy PAPR systems
- High efficiency (HE) respiratory protection against particulates
- 3M Catalog# 450-00-01R12, 453-00-01R06, 453-02-01R06, 453-03-01R06, 453-07-01R06
Specifications
Ammonia, Methylamine Cartridge | |
Magenta/Green | |
Combination Cartridge/Filter | |
453-02-01R06 |
NIOSH, MSHA | |
High Efficiency (HE) | |
Breathe Easy™ PAPR Systems | |
6/Pk. |