3M™ E-A-R™ Classic™ SuperFit™ Ear Plugs
SuperFit™ ear plugs feature a unique ring to help identify how deeply the ear plug is inserted. When minimal to no orange color is visible, the proper insertion is achieved.
Supplier:  3M™ 7000144937
 View more versions of this product
Catalog No. 29-635-115
Explore available promotions
Description
Description
  • Orange fitting ring provides a guideline for proper insertion and quick visual compliance check
  • Conforms to the shape of the ear canal
  • Reusable pillow pack or disposable poly bag packaging available
  • Test compatible with 3M™ E-A-Rfit™ Validation System
Compliance
3M™ ear plugs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Specifications
Specifications
Yes
Large
Yes
Pillow Pk.
Foam
33 dB
Cylinder
No
Class AL
Assembly, Blasting, Cleaning, Demolition, Electrical, Facility Maintenance, Grinding, Machine Operations, Painting, Sanding, Welding
310-1008
2000 Pairs/Cs
Yellow, orange
Uncorded
Single-use