3M™ E-A-R™ Classic™ SuperFit™ Ear Plugs
SuperFit™ ear plugs feature a unique ring to help identify how deeply the ear plug is inserted. When minimal to no orange color is visible, the proper insertion is achieved.
Supplier:  3M™ 7000127172
 View more versions of this product
Catalog No. 29-635-118
Explore available promotions
Description
Description
  • Orange fitting ring provides a guideline for proper insertion and quick visual compliance check
  • Conforms to the shape of the ear canal
  • Reusable pillow pack or disposable poly bag packaging available
  • Test compatible with 3M™ E-A-Rfit™ Validation System
Compliance
3M™ ear plugs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Specifications
Specifications
Yes
Vinyl
Large
Yes
Poly bag
Foam
33 dB
Cylinder
Red
Class AL
Assembly, Blasting, Cleaning, Demolition, Electrical, Facility Maintenance, Grinding, Machine Operations, Painting, Sanding, Welding
311-1125
2000 Pairs/Cs
Yellow, orange
Corded
No