3M™ E-A-R™ Classic™ Plus and Classic Small Ear Plugs
E-A-R™ Classic™ Plus (NRR 33dB) ear plugs are 20% longer than the regular E-A-R Classic ear plug (and the same diameter) to fit deeper into the ear canal and help provide higher noise reduction. ear Classic Small (NRR 29dB) ear plugs are 7% less than that of the Classic ear plug (and the same length) to help provide comfort and protection in smaller ears.
$64.40 - $565.63
Specifications
ANSI S3.1974 Yes
CSA Z94.2 Class AL
For Use With (Application) Assembly, Blasting, Cleaning, Demolition, Electrical, Facility Maintenance, Grinding, Machine Operations, Painting, Sanding, Welding
High Visibility Yes
Pairs per Package 200 Pairs/Pk.
View More Specs
Products 3
Catalog Number Mfr. No. Model Corded Size Packaging Type Price Quantity  
Catalog Number Mfr. No. Model Corded Size Packaging Type Price Quantity  
18-999-4507
View Documents
3M™
7000127207
311-1105 Corded Large Poly bag
Pack of 200 Pair for $136.71
Case of 10 Pack for $225.70
18-999-4505
View Documents
3M™
7000127171
310-1103 Uncorded Small Pillow Pk.
Pack of 200 Pair for $64.40
Case of 10 Pack for $583.10
18-999-4506
View Documents
3M™
7100000262
310-1101 Uncorded Large Pillow Pk.
Pack of 200 Pair for $565.63
Case of 10 Pack for $5,121.30
Description
Description
  • Conforms to the shape of the ear canal to help create a comfortable barrier
  • Slow-recovery ear plug foam is flame-resistant and moisture-resistant
  • Test compatible with 3M™ E-A-Rfit™ Validation System
  • Classic Plus ear plugs are slightly longer than regular E-A-R Classic ear plugs to fit deeper into the ear canal and provide higher noise reduction
  • Classic Small ear plugs are smaller in diameter than regular E-A-R Classic ear plugs to increase comfort in medium and small ear canals while retaining excellent noise attenuation
Compliance
3M™ ear plugs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Specifications
Specifications
Yes
Assembly, Blasting, Cleaning, Demolition, Electrical, Facility Maintenance, Grinding, Machine Operations, Painting, Sanding, Welding
200 Pairs/Pk.
Yellow
Cylinder
No
Class AL
Yes
Foam
33 dB
Single-use
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'