Learn More
3M™ E-A-R™ Push-Ins™ Ear Plugs
A gentle push is all it takes for consistent insertion with E-A-R™ Push-Ins™ ear plugs. Blue stem makes insertion and removal easy and helps keep the tips clean when the wearer’s hands are dirty.
Supplier:  3M™ 7000127213
 View more versions of this product
Catalog No. 19-121-095
Only null left
Explore available promotions
Description
Description
  • Eliminate the need to roll down ear plugs before inserting
  • Mold comfortably to fit different size ear canals
  • E-A-Rform™ soft foam tip provides exceptional comfort and performance
  • Flexible stem for easy insertion and removal
  • Prevent hand-to-ear contamination
Compliance
3M™ ear plugs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Specifications
Specifications
Yes
Universal
No
Poly bag
Foam
28 dB
Pod
No
Class AL
Assembly, Blasting, Cleaning, Demolition, Electrical, Facility Maintenance, Grinding, Machine Operations, Painting, Sanding, Welding
318-1000
400 Pairs/Cs
Yellow
Uncorded
Reusable
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Accessories