missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3M™ E-A-R™ Push-Ins™ Ear Plugs
A gentle push is all it takes for consistent insertion with E-A-R™ Push-Ins™ ear plugs. Blue stem makes insertion and removal easy and helps keep the tips clean when the wearerâs hands are dirty.
$69.63 - $1229.14
Specifications
ANSI S3.1974 | Yes |
---|---|
CSA Z94.2 | Class AL |
Size | Universal |
High Visibility | No |
Packaging Type | Poly bag |
Catalog Number | Mfr. No. | Model | Corded | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Model | Corded | Price | Quantity | |||||
19-121-096
|
3M™
7000052745 |
318-1001 | Corded |
Pack of 100 Pair for $110.28
Case of 4 Pack for $416.24
|
|
|||||
19-121-098
|
3M™
7000052723 |
318-1003 | Corded |
Pack of 200 Pair for $225.90
Case of 10 Pack for $676.00
|
|
|||||
19-121-095
|
3M™
7000127213 |
318-1000 | Uncorded |
Pack of 100 Pair for $69.63
Case of 4 Pack for $262.80
|
|
|||||
19-121-097
|
3M™
7000127185 |
318-1002 | Uncorded |
Case of 2000 Pair for $1,229.14
|
|
|||||
Description
- Eliminate the need to roll down ear plugs before inserting
- Mold comfortably to fit different size ear canals
- E-A-Rform™ soft foam tip provides exceptional comfort and performance
- Flexible stem for easy insertion and removal
- Prevent hand-to-ear contamination
Compliance
3M™ ear plugs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Specifications
Yes | |
Universal | |
Poly bag | |
Yellow | |
Pod | |
No |
Class AL | |
No | |
Foam | |
28 dB | |
Reusable |