3M™ E-A-R™ Push-Ins™ Ear Plugs
A gentle push is all it takes for consistent insertion with E-A-R™ Push-Ins™ ear plugs. Blue stem makes insertion and removal easy and helps keep the tips clean when the wearer’s hands are dirty.
Supplier:  3M™ 7000052723
 View more versions of this product
Catalog No. 19-121-098
Explore available promotions
Description
Description
  • Eliminate the need to roll down ear plugs before inserting
  • Mold comfortably to fit different size ear canals
  • E-A-Rform™ soft foam tip provides exceptional comfort and performance
  • Flexible stem for easy insertion and removal
  • Prevent hand-to-ear contamination
Compliance
3M™ ear plugs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Specifications
Specifications
Yes
Vinyl
Universal
No
Poly bag
Foam
28 dB
Pod
No
Blue
Class AL
Assembly, Blasting, Cleaning, Composite Finishing, Demolition, Electrical, Facility Maintenance, Final Finish, Grinding, Machine Operations, Paint Preparation, Painting, Sanding, Welding
318-1003
2000 Pairs/Cs
Yellow
Corded
Reusable