3M™ PELTOR™ Optime™ 105 Ear Muffs
Optime™ Ear Muffs provide maximum sound quality in the most demanding environments where noise exposures are expected to reach as high as 105 dBA.
Supplier:  3M™ 7000009664
 View more versions of this product
Catalog No. 18-858N
Explore available promotions
Description
Description
  • Effective hearing protection for noise exposures up to 105dBA
  • Double-shell ear cup design technology boosts noise blocking ability
  • Soft, foam-filled ear cushions provide added comfort
  • Durable stainless steel headband resists bending and warping
  • Available in headband style red/black or high-visibility green color
  • Available in neckband and hard hat-attached styles
Compliance
3M™ ear muffs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Specifications
Specifications
Black, Red
Class A
For hard hats
No
1 Ea.
PELTOR™ Optime™
ABS
Yes
Black
Assembly, cleaning, demolition, facility maintenance, grinding, machine operations, painting, sanding, welding
H10B
29 dB
No
Neckband