3M™ PELTOR™ Audio Input Cable
Patch cable that connects certain PELTOR™ headsets to microphones, radios or other personal entertainment devices (listen only). Includes a 36-inch straight cable.
Supplier: 3M™ 7000107805
Description
- Available plug types include 2.5 mm mono, 3.5 mm mono and 2.5 mm stereo
- For use with PELTOR™ headsets that have an available J22 auxiliary port
- Flexible, insulated connection cords made from durable polyurethane
- Meets UL specifications
- Mono plugs also connect to VHF/UFH radios.
- Includes 36-inch straight cable
Compliance
3M™ products must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Specifications
PELTOR™ HT Series Listen-Only Headsets to Connect to Speaker Microphones | |
FL6H | |
Audio Input Cable 3.5 mm mono plug | |
Polyurethane |