missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3M™ PELTOR™ Audio Input Cable
Patch cable that connects certain PELTOR™ headsets to microphones, radios or other personal entertainment devices (listen only). Includes a 36-inch straight cable.
$64.78 - $64.78
Specifications
For Use With (Equipment) | PELTOR™ HT Series Listen-Only Headsets to Connect to Speaker Microphones |
---|---|
Material | Polyurethane |
Catalog Number | Mfr. No. | Model | Product Type | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Model | Product Type | Price | Quantity | |||||
29-636-4189
|
3M™
7000107805 |
FL6H | Audio Input Cable 3.5 mm mono plug |
Each for $64.78
|
|
|||||
Description
- Available plug types include 2.5 mm mono, 3.5 mm mono and 2.5 mm stereo
- For use with PELTOR™ headsets that have an available J22 auxiliary port
- Flexible, insulated connection cords made from durable polyurethane
- Meets UL specifications
- Mono plugs also connect to VHF/UFH radios.
- Includes 36-inch straight cable
Compliance
3M™ products must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Specifications
PELTOR™ HT Series Listen-Only Headsets to Connect to Speaker Microphones | |
Polyurethane |