3M™ PELTOR™ Optime™ 101 Ear Muffs
PELTOR™ Optime™ 101 Ear Muffs are recommended for TWA noise exposures up to 101 dBA.
Supplier:  3M™ 7000009668
 View more versions of this product
Catalog No. 17-388-1B
Explore available promotions
Description
Description
  • For noise exposures up to 101 dBA
  • Liquid-foam ear cushion
  • Ear cushions snap in and out of place for easy cleaning
  • Over-the-Head Style or Hard Hat-Attached styles
  • Adjusts to fit any head size
  • Soft, padded headband for extra comfort
  • Stainless-steel band wires
  • Four-point suspension system distributes suspension evenly
Compliance
3M™ ear muffs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Specifications
Specifications
Black, Green
Liquid-foam
Black
Assembly, cleaning, demolition, facility maintenance, grinding, machine operations, painting, sanding, welding
1 Ea.
PELTOR™ Optime™
Foam, Plastic, Metal
Yes
Class A
For slotted hard hats
H7P3E
24 dB
No
Hard Hat