3M™ PELTOR™ Optime™ 105 Ear Muffs
Optime™ Ear Muffs provide maximum sound quality in the most demanding environments where noise exposures are expected to reach as high as 105 dBA.
Supplier:  3M™ 7000009665
 View more versions of this product
Catalog No. 18-858M
Explore available promotions
Description
Description
  • Effective hearing protection for noise exposures up to 105dBA
  • Double-shell ear cup design technology boosts noise blocking ability
  • Soft, foam-filled ear cushions provide added comfort
  • Durable stainless steel headband resists bending and warping
  • Available in headband style red/black or high-visibility green color
  • Available in neckband and hard hat-attached styles
Compliance
3M™ ear muffs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Specifications
Specifications
Red
Foam
Black
Assembly, cleaning, demolition, facility maintenance, grinding, machine operations, painting, sanding, welding
1 Ea.
PELTOR™ Optime 105
No
Over-the-head
Yes
Class AL
Stainless Steel
H10A
30 dB
Black
ABS