missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3M™ PELTOR™ Optime™ 95 Ear Muffs

PELTOR™ Optime™ 95 Ear Muffs are lightweight and comfortable and protect from noise levels up to 95 dBA.

$33.84 - $365.32

Specifications

Color Beige
ANSI S3.1974 Yes
For Use With (Application) Assembly, cleaning, demolition, facility maintenance, grinding, machine operations, painting, sanding, welding
View More Specs

Products 3
Catalog Number Mfr. No. Model Style Noise Reduction Rating (NRR) Foldable Price Quantity  
Catalog Number Mfr. No. Model Style Noise Reduction Rating (NRR) Foldable Price Quantity  
19-145-412
View Documents
3M™
7000009667
H6A/V Over-the-head 21 dB No
Each for $34.96
Case of 10 Each for $329.30
 
17-388B
View Documents
3M™
7000002328
H6F/V Over-the-head 21 dB Yes
Case of 10 Each for $365.32
 
17-388A
View Documents
3M™
7000009666
H6B/V Behind-the-head 21 dB No
Each for $33.84
Case of 10 Each for $318.70
 
Description

Description

  • Suitable for use when assessed noise levels reach up to 95dBA
  • Slim design for high comfort and continuous wear
  • Behind-the-Head style designed to be worn with most welding helmets
  • Folding style allows convenient storage
  • Over-the-Head style fits well over most safety equipment
  • Beige

Compliance

3M™ ear muffs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.

Specifications

Specifications

Beige
Yes
Assembly, cleaning, demolition, facility maintenance, grinding, machine operations, painting, sanding, welding
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'