missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3M™ Powerflow™ Face-Mounted Powered Air Purifying Respirators

Powered APR provides protection against dust, mist, fumes, radionuclides and radon daughters. Ideal for lead and asbestos.

Supplier:  3M™ 7100010156

 View more versions of this product

Catalog No. 29-635-7472


Explore available promotions

Includes:
  • Medium full facepiece
  • Motor blower unit
  • Battery pack
  • Flow meter

Description

Description

  • High efficiency particulate filter
  • Delivers minimum of 4cfm when used with filter
  • NiCd rechargeable eight-hour battery
  • System uses 6000 Series facepiece
  • Faceshield meets High Impact Requirements of ANSI Z87.1-2003
  • Complete system includes series 6000 full facepiece, 6000DIN, motor blower unit, battery pack, and flow indicator
Specifications

Specifications

ANSI Z87.1-2003
PAPR System with Facepiece
4 CFM
Medium full facepiece, motor blower unit, battery pack, flow meter
Face-mounted
2.131 kg
High Efficiency (HE)
Large
Lead, asbestos, and hazardous particulate
6900PF
4.7 lb.
Videos
SDS
Documents

Documents

Promotions

Promotions

These safety products must be used in accordance with the user instructions, warnings and limitations accompanying each product.