missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3M™ Powerflow™ Face-Mounted Powered Air Purifying Respirators
Powered APR provides protection against dust, mist, fumes, radionuclides and radon daughters. Ideal for lead and asbestos.
$1487.39 - $1487.39
Specifications
ANSI Standard | ANSI Z87.1-2003 |
---|---|
Filter Type | High Efficiency (HE) |
Product Type | PAPR System with Facepiece |
Flow Rate | 4 CFM |
For Use With (Application) | Lead, asbestos, and hazardous particulate |
Includes:
- Medium full facepiece
- Motor blower unit
- Battery pack
- Flow meter
Catalog Number | Mfr. No. | Size | Weight (English) | Weight (Metric) | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Weight (English) | Weight (Metric) | Price | Quantity | |||
29-635-7473
|
3M™
7000052156 |
Medium | 6 lb. | 2.72 kg |
Each for $1,487.39
|
|
|||
29-635-7472
|
3M™
7100010156 |
Large | 4.7 lb. | 2.131 kg |
Each for $1,487.39
|
|
|||
Description
- High efficiency particulate filter
- Delivers minimum of 4cfm when used with filter
- NiCd rechargeable eight-hour battery
- System uses 6000 Series facepiece
- Faceshield meets High Impact Requirements of ANSI Z87.1-2003
- Complete system includes series 6000 full facepiece, 6000DIN, motor blower unit, battery pack, and flow indicator
Specifications
ANSI Z87.1-2003 | |
PAPR System with Facepiece | |
Lead, asbestos, and hazardous particulate | |
Face-mounted |
High Efficiency (HE) | |
4 CFM | |
Medium full facepiece, motor blower unit, battery pack, flow meter |
These safety products must be used in accordance with the user instructions, warnings and limitations accompanying each product.