missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3M™ Powerflow™ Face-Mounted Powered Air Purifying Respirators
Powered APR provides protection against dust, mist, fumes, radionuclides and radon daughters. Ideal for lead and asbestos.
Supplier: 3M™ 7000052156
Includes:
- Medium full facepiece
- Motor blower unit
- Battery pack
- Flow meter
Description
- High efficiency particulate filter
- Delivers minimum of 4cfm when used with filter
- NiCd rechargeable eight-hour battery
- System uses 6000 Series facepiece
- Faceshield meets High Impact Requirements of ANSI Z87.1-2003
- Complete system includes series 6000 full facepiece, 6000DIN, motor blower unit, battery pack, and flow indicator
Specifications
ANSI Z87.1-2003 | |
PAPR System with Facepiece | |
4 CFM | |
Medium full facepiece, motor blower unit, battery pack, flow meter | |
Face-mounted | |
2.72 kg |
High Efficiency (HE) | |
Medium | |
Lead, asbestos, and hazardous particulate | |
6800PF | |
6 lb. |
These safety products must be used in accordance with the user instructions, warnings and limitations accompanying each product.