3M™ Qualitative Fit-Test Kit Accessories
For use with 3M™ Qualitative Fit Test Apparatus
Supplier: 3M™ 7000051788
Description
- Test kits and accessories designed to help your organization meet OSHA fit testing requirements for respirators
- The products can be used to check face-to-respirator seal on any particulate respirator or gas/vapor respirator with a particulate prefilter
- 3M Catalog# 601, FT-13, FT-14, FT-15
Specifications
Respirator Test Hood | |
Used to check face-to-respirator seal on any particulate respirator or gas/vapor respirator with a particulate filter | |
FT-14 |
OSHA | |
Qualitative Fit Test Apparatus FT10 and FT30 | |
5/Cs. |