3M™ Qualitative Fit-Test Kit Accessories
For use with 3M™ Qualitative Fit Test Apparatus
$36.42 - $358.59
Specifications
Certifications/Compliance OSHA
View More Specs
Products 4
Catalog Number Mfr. No. For Use With (Application) Product Type Price Quantity  
Catalog Number Mfr. No. For Use With (Application) Product Type Price Quantity  
19-039434
View Documents
3M™
7000051789
Provide secure hood environment Collar
Case for $158.09
19-039430
View Documents
3M™
7000051788
Used to check face-to-respirator seal on any particulate respirator or gas/vapor respirator with a particulate filter Respirator Test Hood
Pack of 2 Each for $36.42
Case of 5 Pack for $182.10
19-039433
View Documents
3M™
7000001897
Used to check face-to-respirator seal on any particulate respirator or gas/vapor respirator with a particulate filter Nebulizer
Each for $195.31
Case of 3 Each for $585.93
19-080801
View Documents
3M™
7000001945
Quantitative Fit Testing Fit Test Adapter
Each for $358.59
Description
Description
  • Test kits and accessories designed to help your organization meet OSHA fit testing requirements for respirators
  • The products can be used to check face-to-respirator seal on any particulate respirator or gas/vapor respirator with a particulate prefilter
  • 3M Catalog# 601, FT-13, FT-14, FT-15
Specifications
Specifications
OSHA
SDS
missing translation for 'documents'
missing translation for 'documents'