3M™ Qualitative Fit-Test Kit Accessories
For use with 3M™ Qualitative Fit Test Apparatus
Supplier:  3M™ 7000051789
 View more versions of this product
Catalog No. 19-039434
Explore available promotions
Description
Description
  • Test kits and accessories designed to help your organization meet OSHA fit testing requirements for respirators
  • The products can be used to check face-to-respirator seal on any particulate respirator or gas/vapor respirator with a particulate prefilter
  • 3M Catalog# 601, FT-13, FT-14, FT-15
Specifications
Specifications
Collar
Provide secure hood environment
FT-15
OSHA
Qualitative fit test apparatus FT10 and FT30