3M™ Filters and Prefilters for Airstream™ PAPR Systems
Replacement filter parts for use with 3M™ Airstream™ PAPR systems.
Supplier:  3M™ 7100010880
 View more versions of this product
Catalog No. 19-038883
Explore available promotions
Description
Description
  • AS-140 High Efficiency (HE) particulate filter and AS-120-10 prefilter compatible with Airstream™ PAPR systems
  • NIOSH-approved particulate filter provides high efficiency respiratory protection against dust, mist, fumes, asbestos, radionuclides, and radon daughters
  • Prefilter is a component that fits into the PAPR headgear to help filter contaminants before they reach the filter
  • 3M recommends the use of a prefilter for enhanced filtering
  • Easy installation
Specifications
Specifications
NIOSH
HEPA Filter
Airstream™ PAPR Systems
25/Pk.
Magenta
Military, maintenance, mining, oil and gas
AS-140-25