3M™ Filters and Prefilters for Airstream™ PAPR Systems
Replacement filter parts for use with 3M™ Airstream™ PAPR systems.
$85.16 - $1341.84
Specifications
For Use With (Application) | Military, maintenance, mining, oil and gas |
---|---|
For Use With (Equipment) | Airstream™ PAPR Systems |
Description
- AS-140 High Efficiency (HE) particulate filter and AS-120-10 prefilter compatible with Airstream™ PAPR systems
- NIOSH-approved particulate filter provides high efficiency respiratory protection against dust, mist, fumes, asbestos, radionuclides, and radon daughters
- Prefilter is a component that fits into the PAPR headgear to help filter contaminants before they reach the filter
- 3M recommends the use of a prefilter for enhanced filtering
- Easy installation
Specifications
Military, maintenance, mining, oil and gas | |
Airstream™ PAPR Systems |