3M™ Rugged Comfort 6500 Series Half Facepiece Reusable Respirator
Half facepiece respirators are designed to help promote comfort, durability, and stability with a resilient, lightly textured silicone faceseal and strong body construction.
Supplier:  3M™ 7000128240
 View more versions of this product
Catalog No. 19-096-913
Explore available promotions
Description
Description
  • Resilient silicone faceseal offers an extended facepiece life, even keeping its shape in high-heat environments
  • Sleek, low-profile design
  • Special valve cover helps to direct exhaled breath and moisture downward
  • 3M’s proprietary Cool Flow™ Valve helps to make breathing easier by reducing heat and moisture
  • Fits securely and comfortably on the head
  • Includes adjustable head harness assembly featuring long-lasting polyester/spandex straps that stretch and conform to the wearer
  • Highly versatile for many workplace environments and a variety of jobs where respiratory protection is required
  • Compatible with all 3M™ bayonet-style filters and cartridges, including 6000 Series Cartridges and 3M™ 2000, 2200, 7000 or 5000 Series Filters, to help provide respiratory protection against a variety of gases, vapors, and particulate hazards according to NIOSH approvals
Specifications
Specifications
NIOSH 42 CFR Part 84
Bayonet
Half-Mask Respirator
Silicone
Line Compressed Air System SA Series, 3M™ 6000 Series cartridges and 2000, 2200, 7000 or 5000 Series filters
0.37 lb.
OSHA 29 CFR Part 1920.134
Gray, Teal
Large
Facility maintenance, grinding, machine operations, sanding, welding, demolition, assembly, cleaning
6503
0.16 kg