missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3M™ Rugged Comfort 6500 Series Half Facepiece Reusable Respirator
Half facepiece respirators are designed to help promote comfort, durability, and stability with a resilient, lightly textured silicone faceseal and strong body construction.
$38.97 - $38.97
Specifications
Certifications (NIOSH) | NIOSH 42 CFR Part 84 |
---|---|
Certifications (OSHA) | OSHA 29 CFR Part 1920.134 |
Connection Type | Bayonet |
Color | Gray, Teal |
Product Type | Half-Mask Respirator |
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
19-096-911
|
3M™
7000128236 |
Small |
Each for $38.97
Case of 10 Each for $365.00
|
|
|||||
19-096-912
|
3M™
7000128238 |
Medium |
Each for $38.97
Case of 10 Each for $365.00
|
|
|||||
19-096-913
|
3M™
7000128240 |
Large |
Each for $38.97
Case of 10 Each for $365.00
|
|
|||||
Description
- Resilient silicone faceseal offers an extended facepiece life, even keeping its shape in high-heat environments
- Sleek, low-profile design
- Special valve cover helps to direct exhaled breath and moisture downward
- 3Mâs proprietary Cool Flow™ Valve helps to make breathing easier by reducing heat and moisture
- Fits securely and comfortably on the head
- Includes adjustable head harness assembly featuring long-lasting polyester/spandex straps that stretch and conform to the wearer
- Highly versatile for many workplace environments and a variety of jobs where respiratory protection is required
- Compatible with all 3M™ bayonet-style filters and cartridges, including 6000 Series Cartridges and 3M™ 2000, 2200, 7000 or 5000 Series Filters, to help provide respiratory protection against a variety of gases, vapors, and particulate hazards according to NIOSH approvals
Specifications
NIOSH 42 CFR Part 84 | |
Bayonet | |
Half-Mask Respirator | |
Facility maintenance, grinding, machine operations, sanding, welding, demolition, assembly, cleaning | |
0.37 lb. |
OSHA 29 CFR Part 1920.134 | |
Gray, Teal | |
Silicone | |
Line Compressed Air System SA Series, 3M™ 6000 Series cartridges and 2000, 2200, 7000 or 5000 Series filters | |
0.16 kg |