3M™ SecureFit™ Safety Glasses with Anti-fog Lens
Features proprietary 3M™ Pressure Diffusion Temple Technology that helps diffuse pressure over the ear to enhance frame comfort while not compromising the security of fit across a diverse workforce.
Supplier:  3M™ 7100156104
 View more versions of this product
Catalog No. 17-000-712
Explore available promotions
Description
Description
Features:
  • Features proprietary 3M™ Pressure Diffusion Temple Technology which helps diffuse pressure over the ear for a secure, comfortable fit
  • Scotchgard™ Anti-fog Coating lasts longer than traditional anti-fog coatings, helping to increase eyewear performance in wet and steamy environments
  • The proprietary coating retains its effectiveness for at least 25 washings, allowing workers to wear their eyewear longer
  • Meets the requirements of ANSI Z87.1-2015 Standard as an Impact Rated spectacle
  • Polycarbonate lens absorbs 99.9% UVA & UVB rays
  • Scotchgard™ Anti-fog coating provides fog and abrasion resistance.
  • 3M Catalog# SF401SGAF-BLU, SF401SGAF-BLU-F, SF401SGAF-RED, SF402SGAF-BLU, SF402SGAF-BLU-F, SF403SGAF-YEL, SF405SGAF-BLA, SF406SGAF-BLA, SF407SGAF-BLU
Specifications
Specifications
Anti-fog
Removable foam lined gasket helps keep debris out of eyes while providing comfort and cushioning
Safety Glasses
Removable foam lined gasket
20/Pk.
Polycarbonate
UV400
ANSI Z87.1-2015
Clear
Blue, Gray
SF401SGAF-BLU-F
SecureFit™
Plastic
Frameless