3M™ Ultimate FX FF-400 Series Full Facepiece Respirator
Rugged facepiece equipped with a large Scotchgard™ coated lens is designed to provide flexibility in extreme environments.
Supplier:  3M™ 7100001847
 View more versions of this product
Catalog No. 19-177-857
Explore available promotions
Description
Description
  • Large lens with peripheral vision delivers maximum comfort with a wide field of view
  • Exclusive Scotchgard™ coating on lens causes some liquids to bead up on the surface so they can be wiped off easily
  • Durable, permanently attached gaskets for easy maintenance and longer life
  • Low-profile bayonet design improves balance and offers better weight distribution
  • Durable head harness with six-strap configuration fits securely
  • Comfort cradle positions respirator comfortably on head
  • Soft, silicone nose cup and faceseal fits broad range of face shapes and sizes
  • Cool Flow™ exhalation valve allows easier breathing, reduces heat and moisture buildup
Compliance
NIOSH approved under 42CFR84 for use against a variety of respiratory hazards, when used as designed with appropriate cartridges and filters.
Specifications
Specifications
Bayonet
Silicone
Large
Assembly, cleaning, grinding, machine operation, painting, remediation, welding
Speaking diaphragm
FF-403
Black
Full Facepiece Respirator
Silicone
3M™ Particle Filters 2000, 2200, 7000, or 5000 Series, 3M™ 6000 Series Gas/Vapor Cartridges and 3M™ 6000 Combination Cartridges
Scotchgard™
6-point