missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(1,2,3-Thiadiazol-4-yl)benzylamine hydrochloride, ≥95%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC16413CB

 View more versions of this product

Catalog No. CC16413CB


Explore available promotions

Description

Description

Specifications

Specifications

4-(1, 2, 3-Thiadiazol-4-yl)benzylamine hydrochloride
94.5
95%
C9H10ClN3S
250 mg
WUUGGOQPXZDQNW-UHFFFAOYSA-N
[4-(thiadiazol-4-yl)phenyl]methanamine;hydrochloride
2737287
95%
175205-49-9
100.0
Amber Glass Bottle
MFCD02682075
4-1,2,3-thiadiazol-4-yl benzylamine hydrochloride, 4-1,2,3-thiadiazol-4-yl phenyl methanamine hydrochloride, 1-4-1,2,3-thiadiazol-4-yl phenyl methanamine hydrochloride, 4-thiadiazol-4-yl phenyl methanamine hydrochloride, benzenemethanamine,4-1,2,3-thiadiazol-4-yl, 4-1,2,3-thiadiazol-4-yl phenyl methylamine, chloride, 4-4-aminomethyl phenyl-1,2,3-thiadiazole hydrochloride
C1=CC(=CC=C1CN)C2=CSN=N2.Cl
227.71
227.72

Chemical Identifiers

175205-49-9
227.71
WUUGGOQPXZDQNW-UHFFFAOYSA-N
2737287
C1=CC(=CC=C1CN)C2=CSN=N2.Cl
C9H10ClN3S
MFCD02682075
4-1,2,3-thiadiazol-4-yl benzylamine hydrochloride, 4-1,2,3-thiadiazol-4-yl phenyl methanamine hydrochloride, 1-4-1,2,3-thiadiazol-4-yl phenyl methanamine hydrochloride, 4-thiadiazol-4-yl phenyl methanamine hydrochloride, benzenemethanamine,4-1,2,3-thiadiazol-4-yl, 4-1,2,3-thiadiazol-4-yl phenyl methylamine, chloride, 4-4-aminomethyl phenyl-1,2,3-thiadiazole hydrochloride
[4-(thiadiazol-4-yl)phenyl]methanamine;hydrochloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents

Documents

Promotions

Promotions