missing translation for 'onlineSavingsMsg'
Learn More

4-(1-Pyrrolidinyl)benzoic acid, Thermo Scientific™

$30.07 - $30.07

Chemical Identifiers

CAS 22090-27-3
Molecular Formula C11H13NO2
Molecular Weight (g/mol) 191.23
MDL Number MFCD01631241
InChI Key KPCBFFYRSJPCJH-UHFFFAOYSA-N
PubChem CID 2795515
SMILES OC(=O)C1=CC=C(C=C1)N1CCCC1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC30101DA
SDS View Documents
Thermo Scientific Chemicals
CC30101DA
1 g Amber Glass Bottle
Each for $30.07
Only null left
 
Description

Description

Specifications

Chemical Identifiers

22090-27-3
191.23
KPCBFFYRSJPCJH-UHFFFAOYSA-N
OC(=O)C1=CC=C(C=C1)N1CCCC1
C11H13NO2
MFCD01631241
2795515

Specifications

22090-27-3
Amber Glass Bottle
MFCD01631241
4-pyrrolidin-1-yl benzoic acid, 4-1-pyrrolidinyl benzoic acid, 4-pyrrolidin-1-yl-benzoic acid, 4-1-pyrrolidinyl benzenecarboxylic acid, benzoic acid,4-1-pyrrolidinyl, benzoic acid, 4-1-pyrrolidinyl, 4-pyrrolidinylbenzoic acid, pubchem19562, 1-4-carboxyphenyl pyrrolidine
OC(=O)C1=CC=C(C=C1)N1CCCC1
191.23
191.23
4-(1-Pyrrolidinyl)benzoic acid
97%
C11H13NO2
1 g
KPCBFFYRSJPCJH-UHFFFAOYSA-N
4-pyrrolidin-1-ylbenzoic acid
2795515
97%
Videos
Safety and Handling

Safety and Handling

Warning

SDS
missing translation for 'documents'

missing translation for 'documents'