missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
[4-(1H-Pyrrol-1-yl)phenyl]methanol, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC25509DA
Specifications
[4-(1H-Pyrrol-1-yl)phenyl]methanol | |
96.5 | |
97% | |
C11H11NO | |
1g | |
LQQQPLUFBVYLRE-UHFFFAOYSA-N | |
(4-pyrrol-1-ylphenyl)methanol | |
599478 | |
97% |
143426-51-1 | |
100.0 | |
Amber Glass Bottle | |
MFCD03659708 | |
4-1h-pyrrol-1-yl phenyl methanol, 1-4-hydroxymethylphenyl pyrrole, 4-1h-pyrrol-1-yl benzyl alcohol, 4-pyrrol-1-yl phenyl methanol, benzenemethanol,4-1h-pyrrol-1-yl, 4-pyrrolylphenyl methan-1-ol, 4-1h-pyrrol-1-yl benzylalcohol, pubchem11303, 4-pyrrol-1-ylphenyl methanol | |
C1=CN(C=C1)C2=CC=C(C=C2)CO | |
173.215 | |
173.21 |
Chemical Identifiers
143426-51-1 | |
173.215 | |
LQQQPLUFBVYLRE-UHFFFAOYSA-N | |
599478 | |
C1=CN(C=C1)C2=CC=C(C=C2)CO |
C11H11NO | |
MFCD03659708 | |
4-1h-pyrrol-1-yl phenyl methanol, 1-4-hydroxymethylphenyl pyrrole, 4-1h-pyrrol-1-yl benzyl alcohol, 4-pyrrol-1-yl phenyl methanol, benzenemethanol,4-1h-pyrrol-1-yl, 4-pyrrolylphenyl methan-1-ol, 4-1h-pyrrol-1-yl benzylalcohol, pubchem11303, 4-pyrrol-1-ylphenyl methanol | |
(4-pyrrol-1-ylphenyl)methanol |