missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

[4-(1H-Pyrrol-1-yl)phenyl]methanol, 97%, Thermo Scientific™

Chemical Identifiers

CAS 143426-51-1
Molecular Formula C11H11NO
Molecular Weight (g/mol) 173.215
MDL Number MFCD03659708
InChI Key LQQQPLUFBVYLRE-UHFFFAOYSA-N
Synonym 4-1h-pyrrol-1-yl phenyl methanol, 1-4-hydroxymethylphenyl pyrrole, 4-1h-pyrrol-1-yl benzyl alcohol, 4-pyrrol-1-yl phenyl methanol, benzenemethanol,4-1h-pyrrol-1-yl, 4-pyrrolylphenyl methan-1-ol, 4-1h-pyrrol-1-yl benzylalcohol, pubchem11303, 4-pyrrol-1-ylphenyl methanol
PubChem CID 599478
IUPAC Name (4-pyrrol-1-ylphenyl)methanol
SMILES C1=CN(C=C1)C2=CC=C(C=C2)CO
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC25509CB
SDS View Documents
Thermo Scientific Chemicals
CC25509CB
250mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC25509DA
SDS View Documents
Thermo Scientific Chemicals
CC25509DA
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC25509DE
SDS View Documents
Thermo Scientific Chemicals
CC25509DE
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

143426-51-1
173.215
LQQQPLUFBVYLRE-UHFFFAOYSA-N
599478
C1=CN(C=C1)C2=CC=C(C=C2)CO
C11H11NO
MFCD03659708
4-1h-pyrrol-1-yl phenyl methanol, 1-4-hydroxymethylphenyl pyrrole, 4-1h-pyrrol-1-yl benzyl alcohol, 4-pyrrol-1-yl phenyl methanol, benzenemethanol,4-1h-pyrrol-1-yl, 4-pyrrolylphenyl methan-1-ol, 4-1h-pyrrol-1-yl benzylalcohol, pubchem11303, 4-pyrrol-1-ylphenyl methanol
(4-pyrrol-1-ylphenyl)methanol

Specifications

143426-51-1
100.0
Amber Glass Bottle
MFCD03659708
4-1h-pyrrol-1-yl phenyl methanol, 1-4-hydroxymethylphenyl pyrrole, 4-1h-pyrrol-1-yl benzyl alcohol, 4-pyrrol-1-yl phenyl methanol, benzenemethanol,4-1h-pyrrol-1-yl, 4-pyrrolylphenyl methan-1-ol, 4-1h-pyrrol-1-yl benzylalcohol, pubchem11303, 4-pyrrol-1-ylphenyl methanol
C1=CN(C=C1)C2=CC=C(C=C2)CO
173.215
173.21
[4-(1H-Pyrrol-1-yl)phenyl]methanol
96.5
97%
C11H11NO
250mg
LQQQPLUFBVYLRE-UHFFFAOYSA-N
(4-pyrrol-1-ylphenyl)methanol
599478
97%
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'