missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4,4'-Azo-bis(4-cyanovaleric acid), ∽98%, MP Biomedicals™

Supplier:  MP Biomedicals Inc 0215041225

 View more versions of this product

Catalog No. ICN15041225

Explore available promotions

Description

Description

Specifications

Specifications

4, 4'-Azo-bis(4-cyanovaleric acid)
2638-94-0
∽ 98%
MFCD00002799
VFXXTYGQYWRHJP-UHFFFAOYSA-N
4-[(4-carboxy-2-cyanobutan-2-yl)diazenyl]-4-cyanopentanoic acid
280.284
280.28
Polymer synthesis
White
C12H16N4O4
4,4'-azobis 4-cyanovaleric acid, azobis cyanovaleric acid, 4,4'-diazene-1,2-diyl bis 4-cyanopentanoic acid, 4,4'-azobis 4-cyanopentanoic acid, acva, 4,4-azobis 4-cyanovaleric acid, valeric acid, 4,4'-azobis 4-cyano, pentanoic acid, 4,4'-azobis 4-cyano, 4,4'-azobis 4-cyano valeric acid acva
CC(CCC(=O)O)(C#N)N=NC(C)(CCC(=O)O)C#N
25 g
92938
Powder

Chemical Identifiers

2638-94-0
280.284
VFXXTYGQYWRHJP-UHFFFAOYSA-N
92938
CC(CCC(=O)O)(C#N)N=NC(C)(CCC(=O)O)C#N
C12H16N4O4
MFCD00002799
4,4'-azobis 4-cyanovaleric acid, azobis cyanovaleric acid, 4,4'-diazene-1,2-diyl bis 4-cyanopentanoic acid, 4,4'-azobis 4-cyanopentanoic acid, acva, 4,4-azobis 4-cyanovaleric acid, valeric acid, 4,4'-azobis 4-cyano, pentanoic acid, 4,4'-azobis 4-cyano, 4,4'-azobis 4-cyano valeric acid acva
4-[(4-carboxy-2-cyanobutan-2-yl)diazenyl]-4-cyanopentanoic acid
Videos
Safety and Handling

Safety and Handling

storageNote1 : Protect from light.

Recommended Storage : Store at 4°C.

SDS
Documents

Documents

Promotions

Promotions