missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4,4'-Biphenyldisulfonyl Chloride 97.0+%, TCI America™
$157.44 - $408.30
Chemical Identifiers
CAS | 3406-84-6 |
---|---|
Molecular Formula | C12H8Cl2O4S2 |
Molecular Weight (g/mol) | 351.21 |
MDL Number | MFCD00007447 |
InChI Key | OTFAWEIFBPUXOH-UHFFFAOYSA-N |
Synonym | 4,4′C-Biphenyldisulfonyl Dichloride, 4,4′C-Diphenyldisulfonyl Chloride |
PubChem CID | 76950 |
IUPAC Name | [1,1'-biphenyl]-4,4'-disulfonyl dichloride |
SMILES | ClS(=O)(=O)C1=CC=C(C=C1)C1=CC=C(C=C1)S(Cl)(=O)=O |
Chemical Identifiers
3406-84-6 | |
351.21 | |
OTFAWEIFBPUXOH-UHFFFAOYSA-N | |
76950 | |
ClS(=O)(=O)C1=CC=C(C=C1)C1=CC=C(C=C1)S(Cl)(=O)=O |
C12H8Cl2O4S2 | |
MFCD00007447 | |
4,4′C-Biphenyldisulfonyl Dichloride, 4,4′C-Diphenyldisulfonyl Chloride | |
[1,1'-biphenyl]-4,4'-disulfonyl dichloride |
Specifications
3406-84-6 | |
White-Yellow | |
MFCD00007447 | |
3261 | |
OTFAWEIFBPUXOH-UHFFFAOYSA-N | |
[1,1'-biphenyl]-4,4'-disulfonyl dichloride | |
76950 | |
≥97.0% (T) | |
4,4′-Biphenyldisulfonyl Chloride |
207°C | |
C12H8Cl2O4S2 | |
5 g | |
4,4′C-Biphenyldisulfonyl Dichloride, 4,4′C-Diphenyldisulfonyl Chloride | |
ClS(=O)(=O)C1=CC=C(C=C1)C1=CC=C(C=C1)S(Cl)(=O)=O | |
351.21 | |
351.21 | |
Crystalline Powder |
Safety and Handling
TSCA : Yes